4-(6-Chloropyridazin-3-ylmethyl)-1-methanesulfonylazepane

Supplier: Matrix Scientific

068057-250MG
101935-232EA 638.46 USD
101935-232
4-(6-Chloropyridazin-3-ylmethyl)-1-methanesulfonylazepane
4-(6-Chloropyridazin-3-ylmethyl)-1-methanesulfonylazepane

Hazard: Irritant

Formula: C₁₂H₁₈ClN₃O₂S
MDL Number: MFCD19691638
CAS Number: 1316222-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR