1-[4-(5-Bromopyrazin-2-yl)piperidin-1-yl]-2,2-dimethylpropan-1-one
Supplier: Matrix Scientific
068055-250MG
101935-228EA
558.46
USD
101935-228
1-[4-(5-Bromopyrazin-2-yl)piperidin-1-yl]-2,2-dimethylpropan-1-one
1-[4-(5-Bromopyrazin-2-yl)piperidin-1-yl]-2,2-dimethylpropan-1-one
Hazard: Irritant
Formula:
C₁₄H₂₀BrN₃O |
MDL Number:
MFCD19691586 CAS Number: 1316218-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...