3-Chloro-6-(1-methanesulfonylpiperidin-4-ylmethyl)pyridazine

Supplier: Matrix Scientific

068053-250MG
101935-224EA 558.46 USD
101935-224
3-Chloro-6-(1-methanesulfonylpiperidin-4-ylmethyl)pyridazine
3-Chloro-6-(1-methanesulfonylpiperidin-4-ylmethyl)pyridazine

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O₂S
MDL Number: MFCD19691559
CAS Number: 1316221-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR