[5-Iodo-4-(1-methanesulfonylpiperidin-2-yl)pyrimidin-2-yl]dimethylamine

Supplier: Matrix Scientific

068049-250MG
101935-216EA 476.92 USD
101935-216
[5-Iodo-4-(1-methanesulfonylpiperidin-2-yl)pyrimidin-2-yl]dimethylamine
[5-Iodo-4-(1-methanesulfonylpiperidin-2-yl)pyrimidin-2-yl]dimethylamine

Hazard: Irritant

Formula: C₁₂H₁₉IN₄O₂S
MDL Number: MFCD16621983
CAS Number: 1361116-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR