4-(5-Bromo-4-methylthiazol-2-yl)-1-methanesulfonylpiperidine

Supplier: Matrix Scientific

068047-250MG
101935-212EA 476.92 USD
101935-212
4-(5-Bromo-4-methylthiazol-2-yl)-1-methanesulfonylpiperidine
4-(5-Bromo-4-methylthiazol-2-yl)-1-methanesulfonylpiperidine

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂O₂S₂
MDL Number: MFCD16621981
CAS Number: 1361115-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR