[1,4]Oxazepane-6-carboxylic acid methylamide

Supplier: Matrix Scientific

068043-250MG
101935-204EA 638.46 USD
101935-204
[1,4]Oxazepane-6-carboxylic acid methylamide
[1,4]Oxazepane-6-carboxylic acid methylamide

Hazard: Irritant

Formula: C₇H₁₄N₂O₂
MDL Number: MFCD18381750
CAS Number: 1316225-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR