Dimethyl[2-methyl-6-(4-methylpiperidin-4-yl)pyrimidin-4-yl]amine

Supplier: Matrix Scientific

068041-250MG
101935-200EA 687.69 USD
101935-200
Dimethyl[2-methyl-6-(4-methylpiperidin-4-yl)pyrimidin-4-yl]amine
Dimethyl[2-methyl-6-(4-methylpiperidin-4-yl)pyrimidin-4-yl]amine

Hazard: Irritant

Formula: C₁₃H₂₂N₄
MDL Number: MFCD18381787
CAS Number: 1316218-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR