4-Chloro-6-(1-methanesulfonylpiperidin-3-ylmethyl)pyrimidine

Supplier: Matrix Scientific

068039-250MG
101935-196EA 476.92 USD
101935-196
4-Chloro-6-(1-methanesulfonylpiperidin-3-ylmethyl)pyrimidine
4-Chloro-6-(1-methanesulfonylpiperidin-3-ylmethyl)pyrimidine

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O₂S
MDL Number: MFCD19691450
CAS Number: 1316223-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR