4-Chloro-6-(1-methanesulfonylpiperidin-3-yl)-2-methylpyrimidine

Supplier: Matrix Scientific

068037-250MG
101935-192EA 558.46 USD
101935-192
4-Chloro-6-(1-methanesulfonylpiperidin-3-yl)-2-methylpyrimidine
4-Chloro-6-(1-methanesulfonylpiperidin-3-yl)-2-methylpyrimidine

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O₂S
MDL Number: MFCD19691563
CAS Number: 1316218-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR