4-Acetyl-piperazine-2-carboxylic acid dimethylamide hydrochloride
Supplier: Matrix Scientific
068031-500MG
101935-180EA
623.08
USD
101935-180
4-Acetyl-piperazine-2-carboxylic acid dimethylamide hydrochloride
4-Acetyl-piperazine-2-carboxylic acid dimethylamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₈ClN₃O₂ |
MDL Number:
MFCD21606054 CAS Number: 1361111-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...