3-Benzylazetidine-1,3-dicarboxylic acid mono-tert-butyl ester

Supplier: Matrix Scientific

068027-250MG
101935-172EA 553.85 USD
101935-172
3-Benzylazetidine-1,3-dicarboxylic acid mono-tert-butyl ester
3-Benzylazetidine-1,3-dicarboxylic acid mono-tert-butyl ester

Hazard: Irritant

Formula: C₁₆H₂₁NO₄
MDL Number: MFCD21606052
CAS Number: 1361114-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR