1-Benzylpiperazine-2-carboxylic acid amide dihydrochloride

Supplier: Matrix Scientific

068023-500MG
101935-164EA 623.08 USD
101935-164
1-Benzylpiperazine-2-carboxylic acid amide dihydrochloride
1-Benzylpiperazine-2-carboxylic acid amide dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉Cl₂N₃O
MDL Number: MFCD16621933
CAS Number: 1313254-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR