4-(4-Methoxyphenyl)piperazine-2-carboxylic acid methylamide

Supplier: Matrix Scientific

068018-250MG
101935-154EA 476.92 USD
101935-154
4-(4-Methoxyphenyl)piperazine-2-carboxylic acid methylamide
4-(4-Methoxyphenyl)piperazine-2-carboxylic acid methylamide

Hazard: Irritant

Formula: C₁₃H₁₉N₃O₂
MDL Number: MFCD18381527
CAS Number: 1316219-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR