2-Bromo-6-(1-methylpiperidin-3-yl)pyrazine

Supplier: Matrix Scientific

068002-250MG
101935-122EA 558.46 USD
101935-122
2-Bromo-6-(1-methylpiperidin-3-yl)pyrazine
2-Bromo-6-(1-methylpiperidin-3-yl)pyrazine

Hazard: Irritant

Formula: C₁₀H₁₄BrN₃
MDL Number: MFCD19691574
CAS Number: 1316224-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR