2-Pyridin-4-ylpyrrolidine-2-carboxylic acid methylamide

Supplier: Matrix Scientific

067998-250MG
101935-114EA 558.46 USD
101935-114
2-Pyridin-4-ylpyrrolidine-2-carboxylic acid methylamide
2-Pyridin-4-ylpyrrolidine-2-carboxylic acid methylamide

Hazard: Irritant

Formula: C₁₁H₁₅N₃O
MDL Number: MFCD18381637
CAS Number: 1316222-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR