3-[5-(1-Acetylpyrrolidin-2-yl)pyrazin-2-yl]propionic acid

Supplier: Matrix Scientific

067996-250MG
101935-110EA 687.69 USD
101935-110
3-[5-(1-Acetylpyrrolidin-2-yl)pyrazin-2-yl]propionic acid
3-[5-(1-Acetylpyrrolidin-2-yl)pyrazin-2-yl]propionic acid

Hazard: Irritant

Formula: C₁₃H₁₇N₃O₃
MDL Number: MFCD18381781
CAS Number: 1316219-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR