1-Azepan-4-yl-1H-pyrazole-4-carboxylic acid methylamide

Supplier: Matrix Scientific

067985-250MG
101935-088EA 638.46 USD
101935-088
1-Azepan-4-yl-1H-pyrazole-4-carboxylic acid methylamide
1-Azepan-4-yl-1H-pyrazole-4-carboxylic acid methylamide

Hazard: Irritant

Formula: C₁₁H₁₈N₄O
MDL Number: MFCD18381740
CAS Number: 1316221-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR