3-[2-(1-Acetylpiperidin-4-yl)-6-methylpyrimidin-4-yl]propionic acid

Supplier: Matrix Scientific

067983-250MG
101935-084EA 558.46 USD
101935-084
3-[2-(1-Acetylpiperidin-4-yl)-6-methylpyrimidin-4-yl]propionic acid
3-[2-(1-Acetylpiperidin-4-yl)-6-methylpyrimidin-4-yl]propionic acid

Hazard: Irritant

Formula: C₁₅H₂₁N₃O₃
MDL Number: MFCD18381623
CAS Number: 1316220-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR