3-Piperazin-2-ylbenzamide

Supplier: Matrix Scientific

067974-250MG
101935-066EA 638.46 USD
101935-066
3-Piperazin-2-ylbenzamide
3-Piperazin-2-ylbenzamide

Hazard: Irritant

Formula: C₁₁H₁₅N₃O
MDL Number: MFCD18381741
CAS Number: 1316223-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR