1-[2-(3-Chloropyrazin-2-yl)pyrrolidin-1-yl]-2,2-dimethylpropan-1-one

Supplier: Matrix Scientific

067966-250MG
101935-050EA 558.46 USD
101935-050
1-[2-(3-Chloropyrazin-2-yl)pyrrolidin-1-yl]-2,2-dimethylpropan-1-one
1-[2-(3-Chloropyrazin-2-yl)pyrrolidin-1-yl]-2,2-dimethylpropan-1-one

Hazard: Irritant

Formula: C₁₃H₁₈ClN₃O
MDL Number: MFCD19691594
CAS Number: 1316221-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR