2-(5-Bromo-4-methylthiazol-2-yl)pyrrolidine-1-carboxylic acid tert-butyl ester

Supplier: Matrix Scientific

067954-250MG
101935-026EA 558.46 USD
101935-026
2-(5-Bromo-4-methylthiazol-2-yl)pyrrolidine-1-carboxylic acid tert-butyl ester
2-(5-Bromo-4-methylthiazol-2-yl)pyrrolidine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₁₉BrN₂O₂S
MW: 347.28 g/mol
MDL Number: MFCD21606033
CAS Number: 1361113-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR