2-Chloro-6-(1-isopropylpiperidin-2-yl)pyrazine dihydrochloride

Supplier: Matrix Scientific

067952-250MG
101935-022EA 558.46 USD
101935-022
2-Chloro-6-(1-isopropylpiperidin-2-yl)pyrazine dihydrochloride
2-Chloro-6-(1-isopropylpiperidin-2-yl)pyrazine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₀Cl₃N₃
MDL Number: MFCD21606031
CAS Number: 1361114-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR