1-[3-(3-Chloropyrazin-2-ylmethyl)piperidin-1-yl]ethanone

Supplier: Matrix Scientific

067950-250MG
101935-018EA 558.46 USD
101935-018
1-[3-(3-Chloropyrazin-2-ylmethyl)piperidin-1-yl]ethanone
1-[3-(3-Chloropyrazin-2-ylmethyl)piperidin-1-yl]ethanone

Hazard: Irritant

Formula: C₁₂H₁₆ClN₃O
MDL Number: MFCD19691520
CAS Number: 1316227-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR