[5-Iodo-4-(1-methylpiperidin-4-yl)pyrimidin-2-yl]dimethylamine

Supplier: Matrix Scientific

067948-250MG
101935-014EA 476.92 USD
101935-014
[5-Iodo-4-(1-methylpiperidin-4-yl)pyrimidin-2-yl]dimethylamine
[5-Iodo-4-(1-methylpiperidin-4-yl)pyrimidin-2-yl]dimethylamine

Hazard: Irritant

Formula: C₁₂H₁₉IN₄
MDL Number: MFCD16621965
CAS Number: 1361115-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR