2-Chloro-4-(1-methanesulfonylpiperidin-3-ylmethyl)pyridine

Supplier: Matrix Scientific

067936-250MG
101934-990EA 558.46 USD
101934-990
2-Chloro-4-(1-methanesulfonylpiperidin-3-ylmethyl)pyridine
2-Chloro-4-(1-methanesulfonylpiperidin-3-ylmethyl)pyridine

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂S
MDL Number: MFCD19691545
CAS Number: 1316217-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR