1-[2-(2-Dimethylamino-5-iodopyrimidin-4-yl)piperidin-1-yl]ethanone

Supplier: Matrix Scientific

067934-250MG
101934-986EA 476.92 USD
101934-986
1-[2-(2-Dimethylamino-5-iodopyrimidin-4-yl)piperidin-1-yl]ethanone
1-[2-(2-Dimethylamino-5-iodopyrimidin-4-yl)piperidin-1-yl]ethanone

Hazard: Irritant

Formula: C₁₃H₁₉IN₄O
MDL Number: MFCD16621976
CAS Number: 1361114-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR