1-[4-(5-Bromo-4-methylthiazol-2-yl)piperidin-1-yl]ethanone

Supplier: Matrix Scientific

067932-250MG
101934-982EA 476.92 USD
101934-982
1-[4-(5-Bromo-4-methylthiazol-2-yl)piperidin-1-yl]ethanone
1-[4-(5-Bromo-4-methylthiazol-2-yl)piperidin-1-yl]ethanone

Hazard: Irritant

Formula: C₁₁H₁₅BrN₂OS
MDL Number: MFCD16621974
CAS Number: 1361113-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR