6-Bromo-1'-methanesulfonyl-1',2',3',4',5',6'-hexahydro[2,3']bipyridinyl

Supplier: Matrix Scientific

067930-250MG
101934-978EA 558.46 USD
101934-978
6-Bromo-1'-methanesulfonyl-1',2',3',4',5',6'-hexahydro[2,3']bipyridinyl
6-Bromo-1'-methanesulfonyl-1',2',3',4',5',6'-hexahydro[2,3']bipyridinyl

Hazard: Irritant

Formula: C₁₁H₁₅BrN₂O₂S
MW: 319.23 g/mol
MDL Number: MFCD19691537
CAS Number: 1316220-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR