1-[4-(5-Bromopyrazin-2-yl)piperidin-1-yl]ethanone dihydrobromide

Supplier: Matrix Scientific

067911-250MG
101934-940EA 850.77 USD
101934-940
1-[4-(5-Bromopyrazin-2-yl)piperidin-1-yl]ethanone dihydrobromide
1-[4-(5-Bromopyrazin-2-yl)piperidin-1-yl]ethanone dihydrobromide

Hazard: Irritant

Formula: C₁₁H₁₆Br₃N₃O
MDL Number: MFCD21606024
CAS Number: 1361114-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR