1-[4-(3-Chloropyrazin-2-yl)piperidin-1-yl]-2,2-dimethylpropan-1-one

Supplier: Matrix Scientific

067910-250MG
101934-938EA 558.46 USD
101934-938
1-[4-(3-Chloropyrazin-2-yl)piperidin-1-yl]-2,2-dimethylpropan-1-one
1-[4-(3-Chloropyrazin-2-yl)piperidin-1-yl]-2,2-dimethylpropan-1-one

Hazard: Irritant

Formula: C₁₄H₂₀ClN₃O
MDL Number: MFCD19691596
CAS Number: 1316227-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR