4-(3-Chloropyrazin-2-yl)piperidine-1-sulfonic acid dimethylamide

Supplier: Matrix Scientific

067909-250MG
101934-936EA 558.46 USD
101934-936
4-(3-Chloropyrazin-2-yl)piperidine-1-sulfonic acid dimethylamide
4-(3-Chloropyrazin-2-yl)piperidine-1-sulfonic acid dimethylamide

Hazard: Irritant

Formula: C₁₁H₁₇ClN₄O₂S
MDL Number: MFCD21606023
CAS Number: 1361115-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR