1-[4-(4-Chloro-6-methylpyrimidin-2-yl)piperidin-1-yl]ethanone

Supplier: Matrix Scientific

067906-250MG
101934-930EA 558.46 USD
101934-930
1-[4-(4-Chloro-6-methylpyrimidin-2-yl)piperidin-1-yl]ethanone
1-[4-(4-Chloro-6-methylpyrimidin-2-yl)piperidin-1-yl]ethanone

Hazard: Irritant

Formula: C₁₂H₁₆ClN₃O
MDL Number: MFCD19691531
CAS Number: 1316223-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR