1-[4-(6-Chloro-2-methylpyrimidin-4-yl)piperidin-1-yl]ethanone

Supplier: Matrix Scientific

067904-250MG
101934-926EA 558.46 USD
101934-926
1-[4-(6-Chloro-2-methylpyrimidin-4-yl)piperidin-1-yl]ethanone
1-[4-(6-Chloro-2-methylpyrimidin-4-yl)piperidin-1-yl]ethanone

Hazard: Irritant

Formula: C₁₂H₁₆ClN₃O
MDL Number: MFCD19691533
CAS Number: 1316222-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR