4-(6-Chloro-2-methylpyrimidin-4-yl)piperidine-1-sulfonic acid dimethylamide

Supplier: Matrix Scientific

067903-250MG
101934-924EA 558.46 USD
101934-924
4-(6-Chloro-2-methylpyrimidin-4-yl)piperidine-1-sulfonic acid dimethylamide
4-(6-Chloro-2-methylpyrimidin-4-yl)piperidine-1-sulfonic acid dimethylamide

Hazard: Irritant

Formula: C₁₂H₁₉ClN₄O₂S
MDL Number: MFCD21606021
CAS Number: 1361111-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR