(3-Trifluoromethyl-5,6-dihydro-4H-cyclopentapyrazol-1-yl)acetic acid

Supplier: Matrix Scientific

067895-500MG
101934-908EA 320 USD
101934-908
(3-Trifluoromethyl-5,6-dihydro-4H-cyclopentapyrazol-1-yl)acetic acid
(3-Trifluoromethyl-5,6-dihydro-4H-cyclopentapyrazol-1-yl)acetic acid

Hazard: Irritant

Formula: C₉H₉F₃N₂O₂
MDL Number: MFCD02732817
CAS Number: 333309-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR