2-m-Tolylpyrimidine-5-carbaldehyde

Supplier: Matrix Scientific

Synonyms: 2-(3-Methylphenyl)pyrimidine-5-carbaldehyde

067893-500MG
101934-904EA 320 USD
101934-904
2-m-Tolylpyrimidine-5-carbaldehyde
2-m-Tolylpyrimidine-5-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₀N₂O
MW: 198.23 g/mol
MDL Number: MFCD08059844
CAS Number: 915921-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR