3-(2-Fluorophenyl)piperazine-1-carboxylic acid tert-butyl ester
Supplier: Matrix Scientific
067885-250MG
101934-888EA
558.46
USD
101934-888
3-(2-Fluorophenyl)piperazine-1-carboxylic acid tert-butyl ester
3-(2-Fluorophenyl)piperazine-1-carboxylic acid tert-butyl ester
Hazard: Irritant
Formula:
C₁₅H₂₁FN₂O₂ MW: 280.35 g/mol |
MDL Number:
MFCD08685980 CAS Number: 886767-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...