1-Pyrimidin-2-ylpiperidine-2-carboxylic acid

Supplier: Matrix Scientific

067881-250MG
101934-880EA 558.46 USD
101934-880
1-Pyrimidin-2-ylpiperidine-2-carboxylic acid
1-Pyrimidin-2-ylpiperidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₂
MDL Number: MFCD08059687
CAS Number: 1102854-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR