N-(2-Piperidin-4-ylethyl)acetamide dihydrochloride

Supplier: Matrix Scientific

067879-250MG
101934-876EA 558.46 USD
101934-876
N-(2-Piperidin-4-ylethyl)acetamide dihydrochloride
N-(2-Piperidin-4-ylethyl)acetamide dihydrochloride

Hazard: Irritant

Formula: C₉H₂₀Cl₂N₂O
MW: 243.18 g/mol
MDL Number: MFCD21606017
CAS Number: 1361115-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR