1',2',3',4',5',6'-Hexahydro[2,3']bipyridinyl dihydrochloride

Supplier: Matrix Scientific

Synonyms: 2-(Piperidin-3-yl)pyridine dihydrochloride

067876-250MG
101934-870EA 561.54 USD
101934-870
1',2',3',4',5',6'-Hexahydro[2,3']bipyridinyl dihydrochloride
1',2',3',4',5',6'-Hexahydro[2,3']bipyridinyl dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆Cl₂N₂
MW: 235.16 g/mol
MDL Number: MFCD14583121
CAS Number: 51747-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR