4-Methyl-2-piperidin-2-ylpyrimidine-5-carboxylic acid dimethylamide trihydrochloride

Supplier: Matrix Scientific

067856-250MG
101934-830EA 650.77 USD
101934-830
4-Methyl-2-piperidin-2-ylpyrimidine-5-carboxylic acid dimethylamide trihydrochloride
4-Methyl-2-piperidin-2-ylpyrimidine-5-carboxylic acid dimethylamide trihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₃Cl₃N₄O
MW: 357.71 g/mol
MDL Number: MFCD21606012
CAS Number: 1361112-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR