3-[4-(2-Aminopyridin-4-yl)-1H-pyrazol-3-yl]pyrrolidine-1-sulfonic acid dimethylamide

Supplier: Matrix Scientific

067847-250MG
101934-812EA 687.69 USD
101934-812
3-[4-(2-Aminopyridin-4-yl)-1H-pyrazol-3-yl]pyrrolidine-1-sulfonic acid dimethylamide
3-[4-(2-Aminopyridin-4-yl)-1H-pyrazol-3-yl]pyrrolidine-1-sulfonic acid dimethylamide

Hazard: Irritant

Formula: C₁₄H₂₀N₆O₂S
MDL Number: MFCD21606005
CAS Number: 1361114-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR