4-(3'-Fluorobiphenyl-2-ylmethyl)piperidine-4-carboxylic acid isopropylamide

Supplier: Matrix Scientific

067846-250MG
101934-810EA 476.92 USD
101934-810
4-(3'-Fluorobiphenyl-2-ylmethyl)piperidine-4-carboxylic acid isopropylamide
4-(3'-Fluorobiphenyl-2-ylmethyl)piperidine-4-carboxylic acid isopropylamide

Hazard: Irritant

Formula: C₂₂H₂₇FN₂O
MDL Number: MFCD21606004
CAS Number: 1361118-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR