4-(2'-Methylbiphenyl-2-ylmethyl)piperidine-4-carboxylic acid allylamide

Supplier: Matrix Scientific

067845-250MG
101934-808EA 476.92 USD
101934-808
4-(2'-Methylbiphenyl-2-ylmethyl)piperidine-4-carboxylic acid allylamide
4-(2'-Methylbiphenyl-2-ylmethyl)piperidine-4-carboxylic acid allylamide

Hazard: Irritant

Formula: C₂₃H₂₈N₂O
MDL Number: MFCD21606003
CAS Number: 1361114-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR