4-Biphenyl-2-ylmethylpiperidine-4-carboxylic acid propylamide

Supplier: Matrix Scientific

067843-250MG
101934-804EA 476.92 USD
101934-804
4-Biphenyl-2-ylmethylpiperidine-4-carboxylic acid propylamide
4-Biphenyl-2-ylmethylpiperidine-4-carboxylic acid propylamide

Hazard: Irritant

Formula: C₂₂H₂₈N₂O
MW: 336.48 g/mol
MDL Number: MFCD21606001
CAS Number: 1361114-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR