4-(2'-Fluorobiphenyl-4-ylmethyl)piperidine-4-carboxylic acid isopropylamide

Supplier: Matrix Scientific

067841-250MG
101934-800EA 476.92 USD
101934-800
4-(2'-Fluorobiphenyl-4-ylmethyl)piperidine-4-carboxylic acid isopropylamide
4-(2'-Fluorobiphenyl-4-ylmethyl)piperidine-4-carboxylic acid isopropylamide

Hazard: Irritant

Formula: C₂₂H₂₇FN₂O
MDL Number: MFCD21606000
CAS Number: 1361113-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR