2-(2-Methoxyphenyl)-1-(octahydropyrrolo[3,4-b]azepin-1-yl)ethanone hydrochloride

Supplier: Matrix Scientific

067840-250MG
101934-798EA 558.46 USD
101934-798
2-(2-Methoxyphenyl)-1-(octahydropyrrolo[3,4-b]azepin-1-yl)ethanone hydrochloride
2-(2-Methoxyphenyl)-1-(octahydropyrrolo[3,4-b]azepin-1-yl)ethanone hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₅ClN₂O₂
MDL Number: MFCD21605999
CAS Number: 1361114-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR