4-(3-Pyridin-3-ylbenzyl)piperidine-4-carboxylic acid allylamide

Supplier: Matrix Scientific

067839-250MG
101934-796EA 476.92 USD
101934-796
4-(3-Pyridin-3-ylbenzyl)piperidine-4-carboxylic acid allylamide
4-(3-Pyridin-3-ylbenzyl)piperidine-4-carboxylic acid allylamide

Hazard: Irritant

Formula: C₂₁H₂₅N₃O
MDL Number: MFCD21605998
CAS Number: 1361111-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR