Cyclohexylmethylpiperidin-4-ylamine hydrochloride

Supplier: Matrix Scientific

067836-500MG
101934-790EA 380 USD
101934-790
Cyclohexylmethylpiperidin-4-ylamine hydrochloride
Cyclohexylmethylpiperidin-4-ylamine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₅ClN₂
MDL Number: MFCD16621900
CAS Number: 1229627-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR