1-(4-Aminocyclohexyl)piperidin-4-olhydrochloride

Supplier: Matrix Scientific

067830-500MG
101934-778EA 623.08 USD
101934-778
1-(4-Aminocyclohexyl)piperidin-4-olhydrochloride
1-(4-Aminocyclohexyl)piperidin-4-olhydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O
MDL Number: MFCD16621910
CAS Number: 1229625-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR